missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (+/-)-a-Lipoamide, Liquid-Liquid Phase Separated

Product Code. 18749406
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18749406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18749406 Supplier R&D Systems™ Supplier No. 7044/100
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Liquid-liquid phase separated condensate modifier; reduces stress granule formation

(±)-α-Lipoic acid (ALA) is a liquid-liquid phase separated condensate modifier. Reduces stress granule formation in HeLa cells. Also stimulates mitochondrial biogenesis in 3T3-L1 adipocytes. Prevents dieback of FUS mutant motor neurons in culture and reverses motor defects in D. melanogaster expressing mutated human FUS. (±)-α-Lipoic acid is also an antixocidant. In hematopoietic stem cells (HSC) (±)-α-Lipoic acid reduces levels of reactive oxygen species (ROS) and inhibits apoptosis. It also promotes the generation of hematopoietic stem/progenitor cells from hPSCs in vitro and maintenance of functional cord blood HSCs ex vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dithiolane-3-pentanoic acid
CAS 1077-28-7
Quantity 100 mg
Product Line Tocris
Molecular Formula C8H14O2S2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.