missing translation for 'onlineSavingsMsg'
Learn More

a-Methyl-1H-Indazole-4-methanamine, TRC

Product Code. 30403783
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403783 2.5 mg 2.5mg
30396356 5 mg 5mg
30398056 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403783 Supplier Toronto Research Chemicals Supplier No. TRCI5056032.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159511-31-5
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.2
IUPAC Name 1-(1H-indazol-4-yl)ethanamine
SMILES CC(N)c1cccc2[nH]ncc12

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C9H11N3/c1-6(10)7-3-2-4-9-8(7)5-11-12-9/h2-6H,10H2,1H3,(H,11,12)
Formula Weight 161.0953
Chemical Name or Material a-Methyl-1H-Indazole-4-methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.