Learn More
Tuttnauer A032 Petri Dish Insert
Product Code. 17818249 Shop All Tuttnauer Products
Change view
Click to view available options
Quantity:
30 Pk.
Unit Size:
1 unit
Product Code. Quantity unitSize
17818249 30 Pk. 1 unit
1 options
Product Code. 17818249 Supplier Tuttnauer Supplier No. D00285200
Only null left
Add to basket
Add to basket
Insert for petri dishes.
  • A032 Insert for petri dishes.
  • Quantity: 30 pcs.
  • External dimensions W x D x H: 470 x 240 x 77 mm
Specifications
Material Stainless Steel
Product Type Insert
For Use With (Equipment) Tiva Washer
Dimensions (L x W x H) 470 x 240 x 77 mm
Quantity 30 Pk.