Learn More
Tuttnauer A114 Gap Filler
Product Code. 17868269 Shop All Tuttnauer Products
Click to view available options
:
Gap Filler; For use with: Tiva Washer
Product Code. 17868269
Brand: Tuttnauer D00559900
Only null left
Add to basket
Add to basket
This Gap Filler has a hexagon head screw with M8x1 thread to be screwed to the positions of the rack where the nozzles are not placed, to allow the passage of the washing water only in the occupied positions.
  • A114 Hexagon head screw with M8x1 thread To be screwed to the positions of the rack where the Nozzles are not placed, to allow the passage of the washing water only in the occupied positions.
Specifications
Material Stainless Steel
Product Type Nozzle
For Use With (Equipment) Tiva Washer