missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ a1A-AR Degrader 9c

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
α1A-AR Degrader 9c is a selective α1A-adrenergic receptor Degrader (PROTAC®; DC50 = 2.86 μM, 94% α1A-adrenoceptor degradation at 10 μM for 12 h). It comprises the α-adrenoceptor antagonist Prazosin (Cat. No. 0623), joined by a linker to a cereblon ligand Pomalidomide (Cat. No. 6302). α1A-AR Degrader 9c selectively downregulates α1A-adrenoceptor levels, but not α1B-AR and α1D-AR in HEK293 cells. α1A-AR Degrader 9c shows concentration-dependent antiproliferative activity in PC-3 prostate cancer cells (IC50 = 6.12 μM) in vitro, and PC-3 tumor xenografts in vivo.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 2-(2-(2-(2-(4-(4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazine-1-carbonyl)-1 H-1,2,3-triazol-1-yl)ethoxy)ethoxy)ethoxy)-N-(2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)acetamide |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C38H43N11O11 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction