Learn More
Honeywell Safety Products™ A700 Grey TSR Glasses
Product Code. 12930345
Change view
Click to view available options
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
12930345 Each
12940345 Each
11827080 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12930345 Supplier Honeywell Safety Products™ Supplier No. 1015351
Only null left
Add to basket
Edge
Add to basket
Economical and comfortable, these safety glasses are affordably priced and suitable for use in a variety of indoor and outdoor application arenas. Honeywell Safety Products™ A700 Grey TSR Glasses are designed with a sporty style and features for a secure and comfortable fit.
  • General use and protection against impact (45m/s).
  • Well suited for indoor/outdoor applications such as utilities and light manufacturing.
  • Affordably priced and economical.
  • Sporty styling.
  • Secure and comfortable fit.
  • Lightweight design.
  • Soft temple tip pads.
  • Molded universal nose bridge.
  • 8-base polycarbonate, abrasion resistant lenses.
  • 180° field of distortion-free vision.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1549, EC Attestation Number EC1549
Lens Coating Anti-fog, Impact-resistant
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) General use and protection against impact (45m/s). Well suited for indoor / outdoor applications such as utilities and light manufacturing. Building, Construction, Catering, Food, Logistics, Maintenance, Service, Telecom, Transportation and Utility industries.
Frame Color Gray
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Polycarbonate