missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AACOCF3
GREENER_CHOICE

Product Code. 30001257
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30001257

Brand: Tocris Bioscience™ 1462/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phospholipase A2 inhibitor

Inhibitor of cytosolic (85 kDa) phospholipase A2. Also inhibits fatty acid amide hydrolase (FAAH, anandamide amidase) in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149301-79-1
Molecular Formula C21H31F3O

Specifications

Quantity 5 mg
Formula Weight 356.47
Purity >98%
Molecular Weight (g/mol) 356.47
Chemical Name or Material AACOCF3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.