missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ab Buffer Kit

Product Code. 10696495 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10696495 Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10696495 Supplier Cytiva Supplier No. 28903059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Binding buffer (10X stock solution), 50mL; Elution buffer (10X stock solution), 15mL; Neutralizing buffer, 25mL

Premade stock buffers optimized for purification of monoclonal and polyclonal IgG using immobilized Protein A or Protein G

  • Eliminates time-consuming buffer preparation
  • Optimized protocols for Ab SpinTrap™, Protein A HP SpinTrap, Protein G HP SpinTrap columns and HiTrap™ columns prepacked with either Protein A or Protein G media

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ab Buffer kit
For Use With (Application) Used for binding, washing, elution of IgG according to recommended protocols, purification of Monoclonal and Polyclonal IgG using immobilized Protein A or Protein G
For Use With (Equipment) Ab SpinTrap™, Protein A HP SpinTrap, Protein G HP SpinTrap™ columns and HiTrap™ columns
Quantity Each
Includes 50mL Binding Buffer (10X stock solution), 15mL Elution Buffer (10X stock solution), 25mL Neutralizing Buffer
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, trometamol
  • UNKNOWN ACUTE-39.8 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.