Learn More
Tocris Bioscience™ Abacavir hemisulfate
Reverse transcriptase inhibitor; antiretroviral
Brand:  Tocris Bioscience™ 4148/50
Encompass
Product Code. 15706555
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Abacavir hemisulfate is a competitive reverse transcriptase inhibitor; inhibits HIV replication. Orally active and brain penetrant. Antiretroviral agent.
Specifications
Specifications
99%
188062-50-2
50 mg
(1 S,4 R)-4-[2-Amino-6-(cyclopropylamino)-9 H-purin-9-yl]-2-cyclopentene-1-methanol hemisulfate
4°C