missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Abacavir sulfate

Product Code. 16483347
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483347 Supplier MedChemExpress Supplier No. HY17423A
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Abacavir sulfate (Abacavir Hemisulfate) is a competitive, orally active nucleoside reverse transcriptase inhibitor. Abacavir sulfate can inhibits the replication of HIV. Abacavir sulfate shows anticancer activity in prostate cancer cell lines. Abacavir sulfate can trespass the blood-brain-barrier and suppresses telomerase activity.

Specifications

Molecular Weight (g/mol) 433.45 g/mol
Product Type Reference compound
Content And Storage 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Shipping Condition Room temperature
Purity 98.47%
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (115.35 mM∣Need ultrasonic) ∣H2O : 33.33 mg/mL (76.89 mM∣Need ultrasonic)
For Use With (Application) COVID-19-anti-virus
CAS 188062-50-2
Synonym Abacavir Hemisulfate∣ ABC sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.