missing translation for 'onlineSavingsMsg'
Learn More

Abacavir Sulfate, TRC

Product Code. 30132983
Change view
Click to view available options
Quantity:
10 mg
100 mg
5 g
Unit Size:
100mg
10mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132983 5 g 5g
30040409 10 mg 10mg
30040539 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132983 Supplier Toronto Research Chemicals Supplier No. TRCA1050005G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188062-50-2
Molecular Formula 2 C14 H18 N6 O . H2 O4 S
Synonym Abacavir sulfate∣Bis[[(1S,4R)-4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopent-2-enyl]methanol] sulfate∣2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1S,4R)-, sulfate (2:1) (ACI)∣2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1S,4R)-, sulfate (2:1) (salt) (9CI)∣2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1S-cis)-, sulfate (2:1) (salt) (ZCI)∣1592U89 sulfate∣Abacavir hemisulfate∣Abamune∣Abavir∣ABC sulfate∣DRG 0257∣Ziagen

Specifications

Format Neat
Molecular Weight (g/mol) 670.74
Product Type API
Shipping Condition Room Temperature
Quantity 5 g
Chemical Name or Material Abacavir Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.