Learn More
Motic™ Abbe Condenser N.A. 0.90/1.25
Product Code. 15455202
Change view
Click to view available options
For Use With (Equipment):
BA310 and BA410 Series Upright Compound Microscopes
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15455202 BA310 and BA410 Series Upright Compound Microscopes Each
1 options
Product Code. 15455202 Supplier Motic™ Supplier No. 1101001901581
  • ONLINE EXCLUSIVE PRICE
    68.10 €
    60.52 € / Pack of 1
    Save 7.58 €
    11% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Constructed with a high quality plastic surround on metal inner, optic-holding parts. Motic™ Abbe Condenser, N.A. 0.90/1.25 has a 6-blade diaphragm.
  • Abbe-type condenser.
  • High-quality construction (plastic/metal).
  • Includes a 6-blade diaphragm.
  • Includes a slot for sliders.
Specifications
Type Condenser
Description N.A. 0.90/1.25
For Use With (Equipment) BA310 and BA410 Series Upright Compound Microscopes
Includes Six-blade diaphragm, slot for sliders