missing translation for 'onlineSavingsMsg'
Learn More

Abcam 2-Hydroxyhexadecanoic acid, 2-Hydroxy C16:0 fatty acid, 50 mg

Product Code. 32120448 Shop All Abcam Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120448 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120448 Supplier Abcam Supplier No. ab14400850mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody.

  • Storage Buffer: Constituents: 100% 2-Hydroxyhexadecanoic acid
  • Form: Solid

Specifications

Formulation Constituents: 100% 2-Hydroxyhexadecanoic acid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.