missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-ADAM17 (phospho T735) antibody, 100 uL

Product Code. 32057877 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32057877 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32057877 Supplier Abcam Supplier No. ab182630100uL
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal secondary antibody, suitable for WB, with 9 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: WB
  • Species Reactivity: Human
  • Storage Buffer: pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 0.88% Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 0.88% Sodium chloride
Gene Symbols ADAM17
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 100 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Modification State phospho T735
Validated Application abrv WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.