missing translation for 'onlineSavingsMsg'
Learn More

Abcam Anti-EBP50/NHERF-1 antibody, 100 ug

Product Code. 32106455 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32106455 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32106455 Supplier Abcam Supplier No. ab3452100ug
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal secondary antibody, suitable for ICC, IP, WB, IHC-P, with 44 citations.

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ICC, IP, WB, IHC-P, IHC-Fr
  • Species Reactivity: Mouse, Human, Dog and Rat
  • Storage Buffer: Preservative: 0.05% Sodium azide, Constituents: PBS, 0.1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.05% Sodium azide, Constituents: PBS, 0.1% BSA
Gene Symbols NHERF1
Host Species Rabbit
Purification Method Affinity purification Immunogen
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse, Human, Dog and Rat.
Target Species Mouse, Human, Dog, Rat
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Hu, Dog, Rt
Validated Application abrv ICC, IP, WB, IHC-P, IHC-Fr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.