missing translation for 'onlineSavingsMsg'
Learn More

Abcam Arachidonic acid, omega-6 Polyunsaturated fatty acid, 250 mg

Product Code. 32173393 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32173393 250 μg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32173393 Supplier Abcam Supplier No. ab120916250mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, with 5 citations.

  • Storage Buffer: Constituents: 75% Ethanol, 25% Arachidonic acid
  • Form: Liquid

Specifications

Formulation Constituents: 75% Ethanol, 25% Arachidonic acid
Quantity 250 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.