missing translation for 'onlineSavingsMsg'
Learn More

Abcam Autophagy Marker (APG5L/ATG5, ATG16L1, ATG4B, ATG9A, Beclin 1, LC3B) Antibody Sampler Panel, 1 Kit

Product Code. 32093570 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32093570 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32093570 Supplier Abcam Supplier No. ab2285251Kit
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for WB, with 3 citations.

  • Applications: WB
  • Species Reactivity: Mouse, Rat and Human

Specifications

Applications Western Blot
Conjugate Unconjugated
Gene Symbols ATG5
Quantity 1 Kit
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse, Rat and Human.
Target Species Mouse, Rat, Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt, Hu
Validated Application abrv WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.