missing translation for 'onlineSavingsMsg'
Learn More

Abcam Caspase 3/7 Assay Kit (Magic Red), 25 Tests

Product Code. 32116053 Shop All Abcam Products
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116053 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116053 Supplier Abcam Supplier No. ab27077125Test
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for Flow Cyt, Fluorescence Microscopy, Enzyme activity assay, with 4 citations.

  • Applications: Flow Cyt, Fluorescence Microscopy, Enzyme activity assay

Specifications

Product Type Secondary Antibody
Shipping Condition Blue Ice
Conjugate Unconjugated
Quantity 25 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.