missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam ErbB2 / HER2 Antibody Panel (EP1045Y, EPR19547-12, SP3, SP101, CAL27), 1 Kit
Shop All Abcam ProductsDescription
- Species Reactivity: Human
Specifications
Specifications
| Conjugate | Unconjugated |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Test Specificity | This product reacts with Human. |
| Target Species | Human |
| Product Type | Secondary Antibody |
| Shipping Condition | Blue Ice |
| Reactivity abrv | Hu |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?