missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Host Species: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Applications: EIA, WB, IHC
- Species Reactivity: Rabbit
- Storage Buffer: pH: 7.2, Preservative: 0.03% Proclin 300, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 0.5% BSA, 0.15% EDTA
- Form: Liquid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Applications | EIA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | pH: 7.2, Preservative: 0.03% Proclin 300, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 0.5% BSA, 0.15% EDTA |
| Host Species | Goat |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Primary |
| Packaging | Vial |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?