missing translation for 'onlineSavingsMsg'
Learn More

Abcam Phalloidin-iFluor 488 Reagent, 300 Tests

Product Code. 32116088 Shop All Abcam Products
Change view
Click to view available options
Quantity:
300 Tests
Unit Size:
300 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116088 300 Tests 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116088 Supplier Abcam Supplier No. ab176753300Test
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for ICC/IF, Fluorescence Microscopy, with over 300 citations.

  • Applications: ICC/IF, Fluorescence Microscopy
  • Storage Buffer: pH: 7.3, Constituents: 99% Dimethylsulfoxide, 1% Fluorescent-Phalloidin Conjugate
  • Form: Liquid

Specifications

Formulation pH: 7.3, Constituents: 99% Dimethylsulfoxide, 1% Fluorescent-Phalloidin Conjugate
Product Type Secondary Antibody
Shipping Condition Blue Ice
Conjugate Unconjugated
Quantity 300 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.