missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Host Species: Rabbit
- Clonality: Polyclonal
- Applications: ELISA, ICC/IF, IHC-Fr, IHC-P, WB
- Species Reactivity: Rat
- Storage Buffer: Potassium phosphate, Sodium chloride
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Whole Molecule | Yes |
| Test Specificity | This product reacts with Rat. |
| Product Type | Polyclonal Antibody |
| Validated Application abrv | ICC/IF, ELISA, WB, IHC-Fr, IHC-P |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?