missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rabbit monoclonal [M87-3] Anti-Mouse IgG1, IgG2a, IgG2b H&L, 100 uL

Product Code. 32116521 Shop All Abcam Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize':
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
32116521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 32116521 missing translation for 'mfr' Abcam missing translation for 'supplierNo' ab125907100uL
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal secondary antibody, suitable for ELISA, with 5 citations.

  • Host Species: Rabbit
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: ELISA
  • Species Reactivity: Mouse and Rat
  • Storage Buffer: pH: 7.2 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A

Specifications

Applications ELISA
Classification Monoclonal
Clone M87-3
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS
Gene Symbols Ighg1
Host Species Rabbit
Purification Method Affinity purification Protein A
Quantity 100 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse and Rat.
Target Species Mouse, Rat
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt
Validated Application abrv ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.