missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Host Species: Rabbit
- Clonality: Monoclonal
- Isotype: IgG
- Applications: ELISA
- Species Reactivity: Human
- Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 1% BSA
- Form: Liquid
- Purification: Affinity purification Protein A
Specifications
Specifications
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | RM117 |
| Conjugate | Biotin |
| Formulation | Preservative: 0.09% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 1% BSA |
| Gene Symbols | IGHG1 |
| Host Species | Rabbit |
| Purification Method | Affinity purification Protein A |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?