missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat AKI Antibody Array - Membrane (7 Targets), 1X8MEMBRANES

Product Code. 32121113 Shop All Abcam Products
Change view
Click to view available options
Quantity:
8 Membranes
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32121113 8 Membranes Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32121113 Supplier Abcam Supplier No. ab1936541x8Membranes
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for Multiplex Protein Detection.

  • Applications: Multiplex Protein Detection
  • Species Reactivity: Rat

Specifications

Conjugate Unconjugated
Gene Symbols Cst3
Quantity 8 Membranes
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Secondary Antibody
Detection Method Luminescent
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv Multiplex Protein Detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.