missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [11-26] Anti-Mouse IgD delta chain (FITC), 250 ug

Product Code. 32109646 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109646 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109646 Supplier Abcam Supplier No. ab99566250ug
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal antibody, suitable for Flow Cyt, with 2 citations.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ig delta chain C region
Applications Flow Cytometry
Classification Monoclonal
Clone 11-26
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene Ig delta chain C region secreted form
Host Species Rat
Purification Method Affinity purification Protein A
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2a
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv Flow Cyt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.