missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [LO-MM-3] Anti-Mouse IgM heavy chain, 250 ug

Product Code. 32110011 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110011 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110011 Supplier Abcam Supplier No. ab11658250ug
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal antibody, suitable for ELISA.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgM
  • Applications: ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: PBS
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA
Classification Monoclonal
Clone LO-MM-3
Conjugate Unconjugated
Formulation Preservative: 0.09% Sodium azide, Constituents: PBS
Gene Ighm
Gene Symbols Ighm
Host Species Rat
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgM
Host abrv Rt
Shipping Condition Blue Ice
Target Molecule Ighm, Mouse
Validated Application abrv ELISA
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.