missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [SB77e] Anti-Mouse IgG1 H&L (HRP) preadsorbed, 500 uL

Product Code. 32116380 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116380 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116380 Supplier Abcam Supplier No. ab99603500uL
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal secondary antibody, suitable for ELISA, with 5 citations.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Applications: ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
  • Form: Liquid

Specifications

Applications ELISA
Classification Monoclonal
Clone SB77e
Conjugate Unconjugated
Formulation pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine)
Gene Symbols Ighg1
Host Species Rat
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG2b
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.