missing translation for 'onlineSavingsMsg'
Learn More

Abcam VHH Single Domain Anti-Rabbit IgG Fc, 250 uL

Product Code. 32109611 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109611 250 μL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109611 Supplier Abcam Supplier No. ab218695250uL
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Llama Monoclonal antibody, suitable for IHC-P, WB, ELISA, with 24 citations.

  • Host Species: Llama
  • Clonality: Monoclonal
  • Applications: IHC-P, WB, ELISA
  • Species Reactivity: Rabbit
  • Storage Buffer: Constituents: PBS, Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification His Tag

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GD001
Conjugate Unconjugated
Formulation Constituents: PBS, Sodium chloride
Gene Rabbit IgG
Host Species Llama
Purification Method Affinity purification His Tag
Quantity 250 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rabbit.
Target Species Rabbit
Product Type Monoclonal Antibody
Form Liquid
Host abrv Ll
Shipping Condition Blue Ice
Reactivity abrv Rb
Target Molecule Rabbit IgG
Validated Application abrv IHC-P, WB, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.