missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ABI PRISM™ 7000 Sequence Detection Systems Spectral Calibration Kit

Product Code. 10095404 Shop All Applied Biosystems Products
Click to view available options
Quantity:
8 x 96-well plates
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10095404

Brand: Applied Biosystems™ 4328895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the ABI PRISM™ 7000 Sequence Detection System quickly and easily.

This Applied Biosystems™ easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™, ROX™, SYBR™ Green, VIC™, and TAMRA™ dyes on the ABI PRISM™ 7000 Sequence Detection System. The ABI PRISM™ 7000 Sequence Detection Systems Spectral Calibration Kit includes eight optical 96-well reaction plates: one preloaded and sealed background plate, and seven preloaded and sealed spectral calibration plates with seven separate dye standards. Spend Your Time on Research - Not Instrument Calibration This kit's easy-to-use, preloaded, 96-well optical reaction plates enable users to calibrate the ABI PRISM™ 7000 Sequence Detection System quickly and easily. Insert the reaction plates during system installation, and the system automatically collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one Background plate, and seven Spectral Calibration plates with seven dye standards (FAM™, JOE™, NED™,ROX™, SYBR™ Green, VIC™, TAMRA™). Store at -15 to 25°C. Protect from light.
Calibrated Dye FAM, JOE, NED, ROX, TAMRA™, TMR (Tetramethylrhodamine), VIC, SYBR™ Green
For Use With (Equipment) 7000 System
Detection Method Fluorescence
Product Line ABI PRISM
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 8 x 96-well plates
Calibration Type Spectral Calibration

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.