missing translation for 'onlineSavingsMsg'
Learn More

Abietic acid, 85%

Product Code. 10108133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10108133 25 g Glass bottle 25g
10449310 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10108133 Supplier Thermo Scientific Acros Supplier No. 173130250
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 514-10-3 | C20H30O2 | 302.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 514-10-3
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD03423567
InChI Key RSWGJHLUYNHPMX-ONCXSQPRSA-N
Synonym abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
PubChem CID 10569
ChEBI CHEBI:28987
IUPAC Name (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
SMILES CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Brown-Yellow
Assay Percent Range 85%
Beilstein 09, II, 424
Merck Index 15, 7
Formula Weight 302.45
Percent Purity ≥80%
Grade Technical
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystals and Chunks
Chemical Name or Material Abietic acid, 85%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Abietic acid, tech.
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.