missing translation for 'onlineSavingsMsg'
Learn More

Abiraterone Ethyl Ether, TRC

Product Code. 30316835
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316835 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316835 Supplier Toronto Research Chemicals Supplier No. TRCA108525100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abiraterone Ethyl Ether
CAS 2484719-14-2
Purity Grade Notes HPLC
Molecular Formula C26H35NO
Quantity 100 mg
Synonym 3-((3S,8R,9S,10R,13S,14S)-3-ethoxy-10,13-Dimethyl-2,3,4,7,8,9,10,11,12,13,14,15-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl)pyridine
InChI Formula InChI=1S/C26H35NO/c1-4-28-20-11-13-25(2)19(16-20)7-8-21-23-10-9-22(18-6-5-15-27-17-18)26(23,3)14-12-24(21)25/h5-7,9,15,17,20-21,23-24H,4,8,10-14,16H2,1-3H3/t20-,21-,23-,24-,25-,26+/m0/s1
SMILES OCC(O)=O
IUPAC Name 3-[(3S,8R,9S,10R,13S,14S)-3-ethoxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-17-yl]pyridine
Molecular Weight (g/mol) 377.56
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.