missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ABL1 Split FISH Probe
Description
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Format | Tube |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 25 |
| Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Quantity | 200 μL |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?