Learn More
Abnova™ 6q Subtelomere (FITC) FISH Probe
Descrizione
Includes
FISH Hybridization Buffer (80μL)
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifica
Specifica
| Content And Storage | Store at 4°C in the dark. |
| Label or Dye | FITC |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Includes | FISH Hybridization Buffer (80 uL) |
| Location | 6q Subtelomere |
| Product Type | 6q Subtelomere(FITC) FISH Probe |
| Quantity | 20 μL |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.