Learn More
Abnova™ CEN11q (R6G) FISH Probe
Description
Includes
FISH Hybridization Buffer (80μL)
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Label or Dye | R6G |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Includes | FISH Hybridization Buffer (80 uL) |
| Location | 11q12.1 |
| Probe | CEN11q |
| Product Type | CEN11q (R6G) FISH Probe |
| Quantity | 20 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.