missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent In Situ Hybridization (FISH) Buffer
Description
Fluorescent In Situ Hybridization (Cell)
Specifications
Specifications
| Content And Storage | Store at 4°C |
| Description | FISH Hybridization Buffer is optimized for performance with FISH probes and contains dextran sulfate, formamide, and SSC (pH 7.0). |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Product Type | FISH Hybridization Buffer(1ml) |
| Quantity | 1 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?