Learn More
Abnova™ KIF5B/RET SY Translocation FISH Probe
Descripción
Includes
DAPI Counterstain (1500ng/mL ) 250μL.
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Especificaciones
Especificaciones
| Content And Storage | Store at 4°C in the dark. |
| Detection Method | Fluorescence |
| Format | Tube |
| Label or Dye | FITC |
| Final Product Type | Probes (Labeled DNA) |
| Sample Type | Tissue (FFPE) |
| Labeling Target | DNA |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) |
| Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
| Product Type | KIF5B/RET SY Translocation FISH Probe |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.