missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute Blue QPCR Mix, ROX

Product Code. 11862153
Click to view available options
No. of Reactions:
1600 Reactions
400 Reactions
Unit Size:
1600 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 11862153

Brand: Thermo Scientific™ AB4138B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity.

Thermo Scientific™ ABsolute Blue qPCR Mixes contain an inert blue dye to significantly enhance the contrast between reagent and plastic, allowing quick and easy verification that master mix has been dispensed.

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with different ROX passive reference dye concentrations
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 16 x 1.25 mL ABsolute Blue QPCR ROX Mix (2X)
Sufficient for 1600 x 25 μL reactions.

Store at -20°C
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line ABsolute Blue
Product Type qPCR Mix
Quantity 1,600 reactions
Sufficient For 1600 Reactions
Fidelity (vs. Taq) 2 X
No. of Reactions 1600 Reactions
Sample Type DNA, dsDNA
Volume 20 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.