missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Absolute Q™ DNA Digital PCR Master Mix (5X)

Product Code. 17497703 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
200 reactions
Unit Size:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17497703 200 reactions 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17497703 Supplier Applied Biosystems™ Supplier No. A52490
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Applied Biosystems Absolute Q DNA Digital PCR Master Mix (5X) is optimized for use with the QuantStudio Absolute Q Digital PCR System and Applied Biosystems digital PCR (dPCR) assays in a simple workflow with minimal processing steps.

Applied Biosystems Absolute Q DNA Digital PCR Master Mix (5X) is optimized for use with the QuantStudio Absolute Q Digital PCR System and Applied Biosystems digital PCR (dPCR) assays in a simple workflow with minimal processing steps. The 5X formulation enables analysis of higher sample volumes and delivers accurate quantification of DNA targets without using a standard curve.

Features of Absolute Q DNA Digital PCR Master Mix include:
Fast time-to-results—5 minutes of hands-on time and results in ∼90 minutes
Simple workflow—set up the reaction like qPCR, then load and run the plate in a single instrument
High sensitivity—ability to detect down to 0.1% mutant allele frequency
Optimized for multiplexing—detect up to four targets in a single reaction

Simple and efficient workflow
The Absolute Q system simplifies the entire dPCR workflow. Prepare the dPCR reaction mix as you would for qPCR, pipette the reaction mix and isolation buffer into the microfluidic array plate (MAP), and place the plate in the QuantStudio Absolute Q Digital PCR System. Compartmentalization, thermal cycling, and data acquisition of up to four targets occur in a single instrument.

High sensitivity
Absolute Q DNA Digital PCR Master Mix (5X) enables analysis of >95% of the loaded reaction and consistently generates >20,000 microreactions, which maximizes sensitivity. Absolute Q DNA Digital PCR Master Mix (5X) can detect down to 0.1% mutant allele frequency when paired with Absolute Q liquid biopsy dPCR assays.

Maximum specificity and accuracy
Absolute Q DNA Digital PCR Master Mix (5X) is formulated with ROX dye for accurate micro-chamber analysis and with hot-start Taq DNA polymerase and dUTP/UDG to deliver high specificity and carryover contamination control.

DNA methylation analysis
For DNA methylation analysis, use Absolute Q 1-step RT-dPCR Master Mix, 4X (Cat. No. A55146), which does not contain UNG.

Related products
Absolute Q Liquid Biopsy dPCR assays
Absolute Q Viral Titer dPCR assays

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Detection Method Primer-probe
Form Liquid
PCR Method Digital PCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) QuantStudio Absolute Q Digital PCR System
Product Line Absolute Q
Product Type Digital PCR Master Mix
Quantity 200 reactions
Shipping Condition Dry Ice
Sufficient For 200 Reactions
For Use With (Application) Digital PCR (dPCR)
No. of Reactions 200 reactions
Sample Type DNA, cDNA
Volume 360 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.