missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Absolute Q™ Universal DNA Digital PCR Master Mix (5X)

Product Code. 30245081 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245081 Each 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245081 Supplier Applied Biosystems™ Supplier No. A72710
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Absolute Q Universal DNA Digital PCR Master Mix is compatible with a broad range of applications and over 20 million Applied Biosystems predesigned TaqMan Assays, eliminating the need to design custom assays.

Absolute Q Universal DNA Digital PCR Master Mix is compatible with a broad range of applications and over 20 million Applied Biosystems predesigned (and performance-guaranteed*) TaqMan Assays, eliminating the need to design custom assays. With 72 hours of assembled-reaction benchtop stability, this master mix enables increased productivity through automation. Simplify your workflow by avoiding assay design and minimizing assay optimization: the same assay can be used for both qPCR and digital PCR (dPCR). To use this master mix, pair it with Applied Biosystems predesigned TaqMan Assays, MAP16 plates, and the QuantStudio Absolute Q Digital PCR System.

Features of Absolute Q Universal DNA Digital PCR Master Mix include:

  • Universal master mix compatible with a wide range of applications
  • Over 20 million predesigned TaqMan Assays available (with a performance guarantee*), eliminating assay design and enabling flexible target selection
  • 72-hour room temperature stability for convenience and improved productivity through automation
  • Multiplex up to four targets in one reaction, saving time and resources
  • Built-in tolerance for common PCR inhibitors enables robust performance even with challenging samples

A master mix optimized for dPCR performance

Absolute Q Universal DNA Digital PCR Master Mix represents a breakthrough in dPCR technology. This highly optimized master mix is specifically designed to maximize dPCR performance when used with the QuantStudio Absolute Q instrument and MAP16 plates. The master mix expands application possibilities by seamlessly working with predesigned TaqMan Assays. This eliminates the need to design custom assays for your experiments, saving you significant time.

Absolute Q Universal DNA Digital PCR Master Mix has been tested with a range of predesigned TaqMan Assays covering the following applications:

  • Gene expression analysis**
  • Viral vector quantification
  • Mutation detection
  • SNP genotyping
  • Copy number variation analysis

Seamless workflow from qPCR to dPCR

Absolute Q Universal DNA Digital PCR Master Mix offers the convenience of using the same predesigned TaqMan Assay without modifications for both qPCR and dPCR, providing a straightforward and efficient solution for nucleic acid quantification. This minimizes the need for extensive optimization when transitioning your assays to a dPCR format. The performance of TaqMan Assays is guaranteed when used as directed, giving you confidence in your results.

Higher multiplexing and automation

You can further improve the efficiency of your dPCR workflow by multiplexing up to four targets in a single reaction. The 72-hour benchtop stability also enables extended walkaway time and automation. This allows you to achieve a higher throughput with less hands-on time.

Simple and consistent dPCR procedure

Running your experiments with Absolute Q Universal DNA Digital PCR Master Mix is straightforward. Simply select a predesigned TaqMan Assay, combine it with the master mix and your DNA sample, load into the MAP16 plate, and insert the plate into the QuantStudio Absolute Q instrument. The master mix is optimized to work seamlessly with the QuantStudio Absolute Q system to deliver sensitive and precise dPCR results.

Optimized for storage convenience and shelf life

For improved convenience, Absolute Q DNA Digital PCR Universal Master Mix is stored at 4°C, and it has a two-year shelf life from the date of manufacture.

* We guarantee that all our TaqMan Assays will enable you to obtain data you can trust, or we'll replace them at no cost or give you a credit for the assay purchase price.

** We recommend using cDNA generated using SuperScript IV VILO Master Mix for optimal results.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage Store at 2°C to 8°C.
Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance High
PCR Method Digital PCR
Polymerase Taq DNA polymerase
Reaction Speed Fast or Standard
Technique Digital PCR
For Use With (Equipment) QuantStudio™ Absolute Q™ Digital PCR System
Product Type Digital PCR Master Mix
Quantity Each
Shipping Condition Shipped at 2°C to 8°C
Sufficient For 200 Reactions
For Use With (Application) Digital PCR (dPCR), DNA Quantitation, Gene Expression, Genotyping, Microbial Detection
Compatibility Absolute Q™ Digital PCR System
Hot Start Hot Start Taq Polymerase
No. of Reactions 200 Reactions
Reaction Format 9 μL
Sample Type DNA, cDNA
Volume 400 μL
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.