missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Absolute Q™ Viral Titer dPCR Assay - CMV

Product Code. 17447713
Click to view available options
:
Absolute Q Viral Titer dPCR Assay - CMV, 700 Reactions
This item is not returnable. View return policy

Product Code. 17447713

Brand: Applied Biosystems™ A52741

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Absolute Q™ Viral Titer dPCR assays enable easy and accurate quantification of viral vectors.

Applied Biosystems™ Absolute Q™ Viral Titer dPCR assays can be run individually or multiplexed using a custom assay with your target gene of interest to measure concentration and evaluate quality for biopharma and gene therapy research.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Detection Method Digital PCR
Format Tube
PCR Method Digital PCR
Product Line Absolute Q
Compatibility QuantStudio™ Absolute Q™ Digital PCR System
Final Product Type Assay
No. of Reactions 700
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.