Learn More
Tocris Bioscience™ ABT 089 Dihydrochloride
GREENER_CHOICE
High affinity and selective α4β2 nAChR partial agonist
Brand:  Tocris Bioscience™ 5079/10
Encompass
Product Code. 15766436
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
ABT 089 dihydrochloride is a high affinity and selective α4β2 partial agonist (Ki = 16 nM). Exhibits >1000-fold and >10,000-fold selectivity for α4β2 over α1β1γ1 and α7 receptors respectively. Neuroprotective against glutamate-induced toxicity in rat cortical neurons in vitro. Enhances cognitive performance in vivo. Orally bioavailable and brain penetrant.
Specifications
Specifications
ABT 089 dihydrochloride
2-Methyl-3-[(2S)-pyrrolidinylmethoxy]pyridine dihydrochloride
Soluble to 100mM in water and to 20mM in DMSO
Bioactive Small Molecule
161416-61-1
10 mg
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Title
Select an issue
Research Use Only. Not For Human or Veterinary Use