missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ABT 594 Hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ABT 594 hydrochloride is a selective α4β2 nAChR agonist (EC50 = 140 nM for human α4β2 nAChRs, in vitro). Exhibits >900-fold selectivity for α4β2 nAChRs over other neurotransmitter receptors. Displays analgesic properties in various animal models of pain. Potentiates gabapentin mediated analgesia. Also demonstrates cognitive enhancement effects in rodent model of attention deficit.
Specifications
Specifications
| Chemical Name or Material | (R)-5-(Azetidin-2-ylmethoxy)-2-chloropyridine hydrochloride |
| CAS | 203564-54-9 |
| Synonym | Tebanicline |
| Quantity | 10 mg |
| Molecular Formula | C9H11ClN2O.HCL |
| Formula Weight | 235.11 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction