missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABTS ELISA Buffer Kit, PeproTech®

Product Code. 17812035
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17812035 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17812035 Supplier Invitrogen™ Supplier No. 900K00
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

This kit contains the key components required for the analysis of 10 Sandwich ELISA plates. It has been specifically formulated to be used in conjunction with PeproTech’s Standard ELISA Development Kits.

  • Contains the key components required for the analysis of ten sandwich ELISA plates
  • Specifically formulated to be used in conjunction with PeproTech standard ELISA development kits

Included solutions and materials:

  • PBS
  • Wash buffer
  • Blocking buffer
  • Diluent
  • ABTS liquid substrate
  • ELISA microplates
  • Sealing film
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Buffer
Quantity 1 Kit
Detection Method Colorimetric
For Use With (Application) ELISA
Product Line PeproTech
Substrate Type ABTS, HRP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.