missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AC 264613

Product Code. 18025282
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18025282 10 mg 10mg
18004832 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025282 Supplier R&D Systems™ Supplier No. 3370/10
  • ONLINE EXCLUSIVE PRICE
    295.68 €
    266.70 € / 10mg
    Save 28.98 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PAR2 receptor agonist

Potent and selective protease-activated receptor 2 (PAR2) agonist (pEC50 = 7.5). Displays no activity at other PAR subtypes and exhibits no significant activity at over 30 other receptors implicated in nociception and inflammation. Stimulates PI hydrolysis, Ca2+ mobilization and cellular proliferation in vitro (pEC50 values are 6.9, 7.0 and 7.5 respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1051487-82-1
Molecular Formula C19H18BrN3O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 400.27
Purity >98%
Molecular Weight (g/mol) 400.276
Chemical Name or Material AC 264613
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.