missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Ac-Cys-NHMe

Product Code. 30000025
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000025 50 mg 50mg
30000012 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30000025 Supplier R&D Systems™ Supplier No. 7936/50
  • ONLINE EXCLUSIVE PRICE
    651.72 €
    583.80 € / 50mg
    Save 67.92 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Building block for the lysine acylation using conjugating enzymes (LACE) technique

Ac-Cys-NHMe is a non-canonical amino acid that is an essential building block for lysine acylation using conjugating enzymes (LACE) technique. LACE is a chemoenzymatic approach that uses an E2 small ubiquitin-like modifier (SUMO)-conjugating enzyme, Ubc9, to conjugate peptide thioesters containing a C-terminal ubiquitin-derived sequence (LRLRGG) to a lysine residue located in a recognition sequence (IKQE) on a protein via an isopeptide bond. Ac-Cys-NHMe can be reacted with a peptide hydrazide to form a reactive peptide thioester, which is the substrate for Ubc9. LACE enables the site-specific modification of proteins at lysine residues to allow attachment of biochemical probes such as fluorophores to proteins or to engineer proteins for site-specific delivery.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-(Acetylamino)-3-mercapto-N-methylpropanamide
CAS 10061-65-1
Quantity 50 mg
Product Line Tocris
Target Unnatural Amino Acids
Molecular Formula C6H12N2O2S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.