missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ Ac-dC Phosphoramidite, 28-400 finish bottle

Product Code. 17937142 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17937142 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17937142 Supplier Thermo Scientific™ Supplier No. 27202714
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis.

Ac-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. By using Ac-dC, Bz-dA, and DMF-dG monomers, fast cleavage and deprotection can be effected by a 1:1 mixture of aqueous ammonium hydroxide and aqueous methylamine (or AMA) in 10 min. This product is supplied in a 250 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N4-Acetyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)-phosphoramidite
Molecular formula: C41H50N5O8P
Base: dC
Base Protection: Ac
Formula weight: 771.8 g/mol
CAS number: 154110-40-4
HPLC Purity: ≥99%; M-11 Impurity ≤0.01%; No single impurity >0.5%
31P NMR Purity: ≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MerMade Synthesizers
Product Type Ac-dC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxycytidine (dC)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 99%
Quantity 10 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type DNA
Product Line TheraPure
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.