Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Product Code. 30274497 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Revco™ UxF
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series
TSX Series, Revco RLE, HERAfreeze HLE, Forma 89000, TLE Series
TSX Universal
TSX Universal (-ULTS Models)
Thermo Scientific TSU Series
Type:
Access Key Pack (EU) Standalone Cards
Access Key Pack (U.S.) Standalone Cards
Factory installed. No stand-alone orders.
Factory-installed. No stand-alone orders.
Field installed
Field-installed
Unit Size:
Each
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type unitSize
30274497 TSX Universal Field-installed Each
11860553 TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series Access Key Pack (EU) Standalone Cards Pack of 5
11336381 TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series Access Key Pack (U.S.) Standalone Cards Pack of 5
15362600 Thermo Scientific TSU Series Field installed Each
13540770 Revco™ UxF Field installed Each
11336401 TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series Factory installed. No stand-alone orders. Each
15718557 TSX Series, Revco RLE, HERAfreeze HLE, Forma 89000, TLE Series Factory installed. No stand-alone orders. Each
31007818 TSX Universal (-ULTS Models) Factory-installed. No stand-alone orders. -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 30274497 Supplier Thermo Scientific™ Supplier No. FELTSXU
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
  • Card access comes with 5 key cards
  • Field installed - purchased separately
  • Factory installed in the plant with purchase of unit
TRUSTED_SUSTAINABILITY
Specifications
Type Field-installed
For Use With (Equipment) TSX Universal
Includes Card access w/five key cards