missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories for Digital Heating Cooling Drybath

Product Code. 15278894 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Places:
6
12
15
16
20
24
30
35
48
Tube Size:
0.5mL
1.5mL
10mm
12mm
13mm
15mL
15mm
16mm
18mm
20mm
50mL large
50mL small
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places Tube Size unitSize
15278894 12 20mm Each
15258884 48 0.5mL Each
15268884 30 1.5mL Each
15278884 15 15mL Each
15288884 6 50mL small Each
15298884 6 50mL large Each
15208894 35 10mm Each
15218894 24 12mm Each
15228894 24 13mm Each
15248894 20 15mm Each
15258894 16 16mm Each
15268894 12 18mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15278894 Supplier Thermo Scientific™ Supplier No. 88880140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize the performance of our digital heating cooling drybath and digital heating shaking bath by using these accessories.

Use Thermo Scientific™ Accessories for Digital Heating Cooling Bath to optimize flexibility and performance.

  •  Warranty: 2 years
TRUSTED_SUSTAINABILITY

Specifications

Lighting LED
No. of Places 12
Type Block
Tube Size 20mm
Dimensions (L x W x H) 3 x 3.9 x 2 in. (76.5 x 98 x 51 mm)

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.