missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Accessories for Orion™ pH Electrodes
Click to view available options
For Use With (Equipment):
6 mm Electrode, Glass
8 mm Electrode, Glass
Sure-Flow Electrode, Standard Glass Electrode
Type:
Bulb Guard
Electrode Storage Bottle
Unit Size:
Pack of 3
Pack of 5
Beskrivelse
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Storage Bottles
- Can be used with any 6, 8 or 12mm pH electrodes
Electrode Bulb Guards
- Designed for glass body pH electrodes
- Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
Tekniske data
Tekniske data
| Description | Electrode storage bottles |
| For Use With (Equipment) | 8 mm Electrode, Glass |
| Type | Electrode Storage Bottle |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion